Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016607.3 | NP_057691.1 | 5 | 918 | ||
NM_177947.2 | NP_808816.1 | 5 | 890 | |||
NM_177948.2 | NP_808817.1 | 5 | 841 | |||
XM_005262141.2 | XP_005262198.1 | 5 | 1047 | |||
GenBank mRNA | AB039669.1 | - | 5 | 865 | ||
AF211175.1 | - | 1 | 486 | |||
AK222655.1 | - | 5 | 875 | |||
AK291225.1 | - | 5 | 919 | |||
AY359079.1 | - | 1 | 453 | |||
BC005194.1 | - | 5 | 834 | 3932220 | ||
BX647802.1 | - | 5 | 859 | |||
CR933656.1 | - | 5 | 1012 |