Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UIMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199297.2 | NP_001186226.1 | 3 | 487 | ||
NM_001199298.1 | NP_001186227.1 | 2 | 262 | |||
NM_001317961.1 | NP_001304890.1 | 2 | 373 | |||
NM_016290.4 | NP_057374.3 | 2 | 276 | |||
XM_005265930.2 | XP_005265987.1 | 2 | 250 | |||
XM_005265932.2 | XP_005265989.1 | 2 | 229 | |||
XM_005265933.2 | XP_005265990.1 | 2 | 341 | |||
XM_005265934.2 | XP_005265991.1 | 2 | 241 | |||
XM_005265935.2 | XP_005265992.1 | 2 | 250 | |||
XM_005265936.2 | XP_005265993.1 | 2 | 241 | |||
XM_006714871.2 | XP_006714934.1 | 2 | 225 | |||
XM_006714874.2 | XP_006714937.1 | 2 | 225 | |||
XM_011534568.2 | XP_011532870.1 | 2 | 241 | |||
XM_011534570.1 | XP_011532872.1 | 2 | 241 | |||
XM_017009575.1 | XP_016865064.1 | 2 | 250 | |||
XM_017009576.1 | XP_016865065.1 | 2 | 242 | |||
XM_017009577.1 | XP_016865066.1 | 2 | 294 | |||
XM_017009578.1 | XP_016865067.1 | 2 | 250 | |||
XM_017009579.1 | XP_016865068.1 | 2 | 150 | |||
GenBank mRNA | AF349313.1 | - | 2 | 218 | ||
AK023044.1 | - | 3 | 264 | |||
AK304794.1 | - | 2 | 199 | |||
BC006078.1 | - | 2 | 183 | 3659756 | ||
BC032561.1 | - | 2 | 164 | 5578134 | ||
BP270983.1 | - | 2 | 262 | |||
BX537376.1 | - | 2 | 276 | |||
CF994701.1 | - | 2 | 386 |