Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016520.2 | NP_057604.1 | 6 | 481 | ||
XM_011518780.2 | XP_011517082.1 | 5 | 418 | |||
GenBank mRNA | AF151054.1 | - | 6 | 472 | ||
AF218421.1 | - | 6 | 436 | |||
AK057004.1 | - | 6 | 463 | |||
AK097577.1 | - | 3 | 1243 | |||
AL137549.1 | - | 4 | 267 | |||
AY632862.1 | - | 3 | 172 | |||
BC007664.2 | - | 6 | 434 | 3140172 |