Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIPPLY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317768.1 | NP_001304697.1 | 4 | 537 | ||
NM_001317777.1 | NP_001304706.1 | 3 | 417 | |||
NM_018962.2 | NP_061835.1 | 4 | 767 | |||
GenBank mRNA | AB037158.1 | - | 4 | 767 | ||
AB037159.1 | - | 4 | 537 | |||
AB037160.1 | - | 3 | 417 | |||
AB037161.1 | - | 1 | 403 |