Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC37A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320537.1 | NP_001307466.1 | 2 | 1072 | ||
NM_018964.3 | NP_061837.3 | 3 | 441 | |||
XM_017028377.1 | XP_016883866.1 | 3 | 366 | |||
XM_017028378.1 | XP_016883867.1 | 3 | 825 | |||
XM_017028379.1 | XP_016883868.1 | 4 | 472 | |||
XM_017028380.1 | XP_016883869.1 | 2 | 272 | |||
XM_017028381.1 | XP_016883870.1 | 3 | 366 | |||
XM_017028383.1 | XP_016883872.1 | 3 | 297 | |||
GenBank mRNA | AF311320.1 | - | 2 | 1011 | ||
AJ269529.2 | - | 3 | 441 | |||
AJ277912.1 | - | 3 | 541 | |||
AJ277913.1 | - | 3 | 467 | |||
BC042139.1 | - | 3 | 327 | |||
DA755019.1 | - | 3 | 366 |