Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018982.4 | NP_061855.1 | 7 | 828 | ||
NR_036639.1 | 7 | 828 | ||||
NR_036640.1 | 6 | 608 | ||||
NR_135075.1 | 5 | 545 | ||||
XM_017001499.1 | XP_016856988.1 | 6 | 586 | |||
XM_017001500.1 | XP_016856989.1 | 6 | 649 | |||
XM_017001501.1 | XP_016856990.1 | 5 | 430 | |||
GenBank mRNA | AK300240.1 | - | 6 | 690 | ||
AK315183.1 | - | 7 | 806 | |||
AL713668.1 | - | 6 | 539 | |||
BC009674.1 | - | 2 | 141 | |||
BC064590.1 | - | 7 | 794 | 5749036 | ||
BX405963.2 | - | 7 | 820 |