Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018984.3 | NP_061857.3 | 15 | 2458 | ||
XM_005268984.4 | XP_005269041.1 | 15 | 3169 | |||
XM_005268985.2 | XP_005269042.1 | 14 | 2284 | |||
XM_011538497.1 | XP_011536799.1 | 14 | 2550 | |||
XM_011538499.1 | XP_011536801.1 | 13 | 2349 | |||
XM_011538500.2 | XP_011536802.1 | 14 | 2374 | |||
XM_011538501.2 | XP_011536803.1 | 14 | 2370 | |||
XM_017019491.1 | XP_016874980.1 | 14 | 3396 | |||
XM_017019492.1 | XP_016874981.1 | 15 | 2511 | |||
XM_017019493.1 | XP_016874982.1 | 13 | 2255 | |||
XM_017019494.1 | XP_016874983.1 | 13 | 2350 | |||
GenBank mRNA | AB037719.1 | - | 5 | 1493 | ||
AB072355.1 | - | 15 | 2371 | |||
BC062341.1 | - | 15 | 2450 | 6376684 |