Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SASH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018990.3 | NP_061863.1 | 6 | 787 | ||
XM_006724763.1 | XP_006724826.1 | 5 | 904 | |||
GenBank mRNA | AK292023.1 | - | 6 | 735 | ||
AK296665.1 | - | 5 | 839 | |||
AK314869.1 | - | 6 | 754 | |||
AL049683.1 | - | 6 | 712 | |||
BC051881.1 | - | 6 | 719 | 6011968 |