Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284301.2 | NP_001271230.1 | 4 | 837 | ||
NM_001284302.2 | NP_001271231.1 | 4 | 837 | |||
NM_018685.4 | NP_061155.2 | 4 | 837 | |||
XM_006715746.1 | XP_006715809.1 | 4 | 834 | |||
XM_006715747.3 | XP_006715810.1 | 4 | 834 | |||
XM_017012354.1 | XP_016867843.1 | 4 | 834 | |||
XM_017012355.1 | XP_016867844.1 | 4 | 834 | |||
XM_017012356.1 | XP_016867845.1 | 4 | 834 | |||
GenBank mRNA | AK291254.1 | - | 4 | 769 | ||
AK299803.1 | - | 4 | 539 | |||
BC070066.1 | - | 4 | 819 | 4825802 |