Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304564.1 | NP_001291493.1 | 6 | 1404 | ||
NM_019008.5 | NP_061881.2 | 6 | 1454 | |||
NR_130789.1 | 6 | 1368 | ||||
NR_130790.1 | 7 | 1518 | ||||
XM_011530236.1 | XP_011528538.1 | 8 | 1522 | |||
XM_011530237.1 | XP_011528539.1 | 7 | 1642 | |||
XM_011530238.1 | XP_011528540.1 | 7 | 1726 | |||
XM_017028838.1 | XP_016884327.1 | 7 | 1620 | |||
GenBank mRNA | AK000239.1 | - | 3 | 625 | ||
AK290954.1 | - | 7 | 1351 | |||
AL136768.1 | - | 6 | 1404 | |||
AL365515.1 | - | 6 | 1331 | |||
AL834205.1 | - | 6 | 1296 | |||
BC002587.2 | - | 2 | 596 | |||
BC008327.1 | - | 6 | 1308 | 3502711 |