Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304564.1 | NP_001291493.1 | 5 | 976 | ||
NM_019008.5 | NP_061881.2 | 5 | 1026 | |||
NR_130789.1 | 5 | 940 | ||||
NR_130790.1 | 6 | 1090 | ||||
XM_011530236.1 | XP_011528538.1 | 7 | 1094 | |||
XM_011530237.1 | XP_011528539.1 | 6 | 1214 | |||
XM_011530238.1 | XP_011528540.1 | 6 | 1298 | |||
XM_017028838.1 | XP_016884327.1 | 6 | 1192 | |||
GenBank mRNA | AK000239.1 | - | 2 | 197 | ||
AK290954.1 | - | 6 | 923 | |||
AL136768.1 | - | 5 | 976 | |||
AL365515.1 | - | 5 | 903 | |||
AL834205.1 | - | 5 | 868 | |||
BC002587.2 | - | 1 | 168 | |||
BC008327.1 | - | 5 | 880 | 3502711 |