Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190326.1 | NP_001177255.1 | 12 | 2535 | ||
NM_022720.6 | NP_073557.3 | 13 | 2634 | |||
XM_006724268.2 | XP_006724331.1 | 10 | 1269 | |||
GenBank mRNA | AK025427.1 | - | 1 | 73 | ||
AK025539.1 | - | 12 | 1598 | |||
AK025780.1 | - | 8 | 910 | |||
AK094381.1 | - | 1 | 1424 | |||
AK222796.1 | - | 12 | 1598 | |||
AY189282.1 | - | 9 | 928 | |||
BC009323.2 | - | 11 | 1437 | |||
BC037564.1 | - | 13 | 2584 | |||
BC078147.1 | - | 11 | 2249 | 6014346 | ||
BX649187.1 | - | 10 | 1526 | |||
CR456356.1 | - | 12 | 2292 |