Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019037.2 | NP_061910.1 | 2 | 468 | ||
XM_011517134.2 | XP_011515436.1 | 2 | 326 | |||
GenBank mRNA | AF281133.1 | - | 2 | 461 | ||
AK000598.1 | - | 2 | 385 | |||
BC002777.2 | - | 2 | 382 | 3613853 | ||
BM839994.1 | - | 2 | 468 |