Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040446.2 | NP_001035536.1 | 3 | 435 | ||
NM_001294343.1 | NP_001281272.1 | 3 | 435 | |||
NM_001294344.1 | NP_001281273.1 | 3 | 435 | |||
XM_005248313.3 | XP_005248370.1 | 3 | 399 | |||
XM_011514057.1 | XP_011512359.1 | 3 | 399 | |||
XM_011514058.2 | XP_011512360.1 | 3 | 399 | |||
XM_011514059.1 | XP_011512361.1 | 3 | 399 | |||
XM_011514060.1 | XP_011512362.1 | 3 | 399 | |||
XM_017009589.1 | XP_016865078.1 | 3 | 399 | |||
XM_017009590.1 | XP_016865079.1 | 4 | 337 | |||
XM_017009591.1 | XP_016865080.1 | 2 | 348 | |||
XM_017009592.1 | XP_016865081.1 | 2 | 212 | |||
XM_017009593.1 | XP_016865082.1 | 3 | 399 | |||
XM_017009594.1 | XP_016865083.1 | 3 | 415 | |||
XM_017009595.1 | XP_016865084.1 | 2 | 342 | |||
XM_017009596.1 | XP_016865085.1 | 3 | 351 | |||
GenBank mRNA | AB051469.1 | - | 3 | 368 | ||
AK000483.1 | - | 3 | 396 | |||
AY028703.1 | - | 3 | 372 | |||
BC057393.1 | - | 3 | 365 | 6012452 | ||
DB483084.1 | - | 3 | 435 |