Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144952.1 | NP_001138424.1 | 43 | 5938 | ||
XM_011524914.2 | XP_011523216.1 | 42 | 6265 | |||
XM_011524915.2 | XP_011523217.1 | 43 | 6324 | |||
GenBank mRNA | AK001694.1 | - | 10 | 1325 | ||
AK091827.1 | - | 3 | 936 | |||
AL137357.1 | - | 21 | 2846 | |||
BC045763.1 | - | 24 | 3409 | |||
BC066363.1 | - | 35 | 4838 |