Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276378.1 | NP_001263307.1 | 7, 8 | 897 | ||
NM_001276379.1 | NP_001263308.1 | 6, 7 | 772 | |||
NM_020347.3 | NP_065080.1 | 5, 6 | 616 | |||
NR_075080.1 | 4, 5 | 491 | ||||
XM_006713207.2 | XP_006713270.2 | 5, 6 | 601 | |||
XM_011533838.2 | XP_011532140.1 | 5, 6 | 540 | |||
XM_017006644.1 | XP_016862133.1 | 8, 9 | 1254 | |||
XM_017006645.1 | XP_016862134.1 | 7, 8 | 1129 | |||
GenBank mRNA | AJ297351.1 | - | 5, 6 | 573 | ||
AK093705.1 | - | 7, 8 | 1215 | |||
AK303416.1 | - | 8, 9 | 668 | |||
BC042483.1 | - | 5, 6 | 576 | 4831113 | ||
BX640604.1 | - | 5, 6 | 601 | |||
CN429018.1 | - | 4, 5 | 401 | |||
DA408809.1 | - | 4, 5 | 462 |