Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099784.2 | NP_001093254.2 | 8 | 1534 | ||
NM_001282351.1 | NP_001269280.1 | 8 | 1402 | |||
GenBank mRNA | AK098777.1 | - | 6 | 1063 | ||
AK127701.1 | - | 8 | 1386 | |||
AK302669.1 | - | 8 | 1536 | |||
BC059173.1 | - | 7 | 1050 | 5738074 |