Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM63B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040450.2 | NP_001035540.1 | 1 | 731 | ||
NM_001040453.2 | NP_001035543.1 | 1 | 561 | |||
XM_011521687.1 | XP_011519989.1 | 1 | 729 | |||
XM_011521688.1 | XP_011519990.1 | 1 | 729 | |||
XM_011521689.1 | XP_011519991.1 | 1 | 729 | |||
XM_011521690.1 | XP_011519992.1 | 1 | 729 | |||
XM_017022332.1 | XP_016877821.1 | 1 | 729 | |||
XM_017022333.1 | XP_016877822.1 | 1 | 729 | |||
XM_017022334.1 | XP_016877823.1 | 1 | 729 | |||
XM_017022335.1 | XP_016877824.1 | 1 | 729 | |||
GenBank mRNA | AB075509.1 | - | 1 | 606 | ||
AK075319.1 | - | 1 | 609 | |||
AK297291.1 | - | 1 | 609 | |||
AL833610.1 | - | 1 | 732 | |||
BC140811.1 | - | 1 | 561 | 9021679 | ||
BC144616.1 | - | 1 | 561 | 9053147 |