Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIT1 SIRNA Assays ›
Gene Symbol:
TRIT1
Gene Name:
tRNA isopentenyltransferase 1
Gene Aliases:
GRO1, IPPT, IPT, IPTase, MOD5, hGRO1
Chromosome Location:
Chr.1: 39841031 - 39883511 on Build GRCh38
UniGene ID:
Hs.356554
Species:
Human
siRNA ID
HSS123277
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001312691.1
NP_001299620.1
9
1071
NM_001312692.1
NP_001299621.1
8
903
NM_017646.5
NP_060116.2
10
1149
NR_132401.1
10
1145
NR_132402.1
9
1003
NR_132403.1
9
999
NR_132404.1
9
999
NR_132405.1
9
995
NR_132406.1
8
886
NR_132407.1
7
763
NR_132408.1
7
759
NR_132409.1
6
620
NR_132410.1
6
646
NR_132412.1
5
507
NR_132413.1
3
317
NR_132415.1
10
1236
XM_005270954.2
XP_005271011.1
10
1385
XM_006710706.1
XP_006710769.1
10
1217
GenBank mRNA
AF074918.1
-
10
1230
AK000068.1
-
8
766
AK074222.1
-
8
766
AK223478.1
-
10
1142
AK307910.1
-
10
1135
AY052768.1
-
9
1033
AY303390.1
-
10
1143
AY702933.1
-
10
1072
AY702934.1
-
10
1068
AY702935.1
-
9
919
AY702936.1
-
9
926
AY702937.1
-
9
922
AY702938.1
-
9
922
AY702939.1
-
9
918
AY702940.1
-
8
809
AY702941.1
-
7
686
AY702942.1
-
7
682
AY702943.1
-
6
543
AY702944.1
-
6
569
AY702945.1
-
5
430
AY702946.1
-
3
240
BC010741.1
-
8
865
BC107569.1
-
10
1148
4594069
BC128155.1
-
10
1143
40112695
BC128156.1
-
10
1230
40112698
CR457226.1
-
7
700
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA processing factor
Gene Ontology Categories:
Function(s)
Process(es)
tRNA modification
mitochondrial tRNA modification
nucleic acid binding
ATP binding
zinc ion binding
tRNA dimethylallyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0