Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312692.1 | NP_001299621.1 | 6 | 736 | ||
NM_017646.5 | NP_060116.2 | 8 | 982 | |||
NR_132401.1 | 8 | 978 | ||||
NR_132402.1 | 7 | 836 | ||||
NR_132403.1 | 7 | 832 | ||||
NR_132404.1 | 7 | 832 | ||||
NR_132405.1 | 7 | 828 | ||||
NR_132406.1 | 6 | 719 | ||||
NR_132407.1 | 5 | 596 | ||||
NR_132408.1 | 5 | 592 | ||||
NR_132409.1 | 4 | 453 | ||||
NR_132410.1 | 4 | 479 | ||||
NR_132412.1 | 3 | 340 | ||||
NR_132415.1 | 8 | 1069 | ||||
XM_005270954.2 | XP_005271011.1 | 8 | 1218 | |||
XM_006710706.1 | XP_006710769.1 | 8 | 1050 | |||
GenBank mRNA | AF074918.1 | - | 8 | 1063 | ||
AK000068.1 | - | 6 | 599 | |||
AK074222.1 | - | 6 | 599 | |||
AK223478.1 | - | 8 | 975 | |||
AK307910.1 | - | 8 | 967 | |||
AY303390.1 | - | 8 | 976 | |||
AY702933.1 | - | 8 | 905 | |||
AY702934.1 | - | 8 | 901 | |||
AY702935.1 | - | 7 | 752 | |||
AY702936.1 | - | 7 | 759 | |||
AY702937.1 | - | 7 | 755 | |||
AY702938.1 | - | 7 | 755 | |||
AY702939.1 | - | 7 | 751 | |||
AY702940.1 | - | 6 | 642 | |||
AY702941.1 | - | 5 | 519 | |||
AY702942.1 | - | 5 | 515 | |||
AY702943.1 | - | 4 | 376 | |||
AY702944.1 | - | 4 | 402 | |||
AY702945.1 | - | 3 | 263 | |||
BC010741.1 | - | 6 | 698 | |||
BC107569.1 | - | 8 | 981 | 4594069 | ||
BC128155.1 | - | 8 | 976 | 40112695 | ||
BC128156.1 | - | 8 | 1063 | 40112698 | ||
BE294565.1 | - | 5 | 600 | |||
CR457226.1 | - | 5 | 533 |