Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018088.2 | NP_001018098.1 | 42 | 4831 | ||
NM_017684.4 | NP_060154.3 | 40 | 4702 | |||
NM_018080.3 | NP_060550.2 | 40 | 4702 | |||
NM_020821.2 | NP_065872.1 | 42 | 4831 | |||
XM_011521713.2 | XP_011520015.1 | 42 | 4792 | |||
XM_011521714.2 | XP_011520016.1 | 42 | 4792 | |||
GenBank mRNA | AJ608770.1 | - | 40 | 4685 | ||
AJ608771.1 | - | 42 | 4814 | |||
AJ626860.1 | - | 40 | 4685 | |||
AJ626861.1 | - | 42 | 4814 |