Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CC2D1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017721.4 | NP_060191.3 | 7, 8 | 1164 | ||
XM_005259972.2 | XP_005260029.1 | 7, 8 | 1112 | |||
XM_005259973.2 | XP_005260030.1 | 7, 8 | 1112 | |||
XM_005259974.2 | XP_005260031.1 | 7, 8 | 1112 | |||
XM_005259975.2 | XP_005260032.1 | 7, 8 | 1112 | |||
GenBank mRNA | AB097002.1 | - | 7, 8 | 1106 | ||
AF536205.1 | - | 9, 10 | 1217 | |||
AK000248.1 | - | 923 | ||||
AK123155.1 | - | 5, 6 | 922 | |||
AK315068.1 | - | 7, 8 | 1082 | |||
BC006556.2 | - | 3, 4 | 402 | |||
BC064981.1 | - | 7, 8 | 1068 | 6585236 |