Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPP8 SIRNA Assays ›
Gene Symbol:
DPP8
Gene Name:
dipeptidyl peptidase 8
Gene Aliases:
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141
Chromosome Location:
Chr.15: 65442463 - 65517713 on Build GRCh38
UniGene ID:
Hs.458609
Species:
Human
siRNA ID
HSS123433
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPP8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320875.1
NP_001307804.1
9, 10
1397
NM_001320876.1
NP_001307805.1
8, 9
1366
NM_017743.5
NP_060213.2
8, 9
1261
NM_130434.4
NP_569118.1
8, 9
1261
NM_197960.3
NP_932064.1
9, 10
1302
NM_197961.3
NP_932065.1
9, 10
1397
NR_135485.1
8, 9
1261
NR_135486.1
9, 10
1302
XM_011521730.1
XP_011520032.1
7, 8
2521
XM_011521731.1
XP_011520033.1
7, 8
2476
XM_011521732.2
XP_011520034.1
8, 9
2650
XM_011521733.1
XP_011520035.1
7, 8
1068
XM_011521734.1
XP_011520036.1
8, 9
2650
XM_017022373.1
XP_016877862.1
8, 9
2650
XM_017022374.1
XP_016877863.1
8, 9
2650
XM_017022375.1
XP_016877864.1
7, 8
1113
XM_017022376.1
XP_016877865.1
8, 9
1242
XM_017022377.1
XP_016877866.1
6, 7
939
XM_017022378.1
XP_016877867.1
8, 9
2650
XM_017022379.1
XP_016877868.1
7, 8
1068
XM_017022380.1
XP_016877869.1
7, 8
1068
XM_017022381.1
XP_016877870.1
6, 7
939
GenBank mRNA
AF221634.1
-
8, 9
1235
AF221636.1
-
1, 2
72
AK000290.1
-
8, 9
1184
AK027826.1
-
4, 5
440
AK291057.1
-
9, 10
1372
AK296915.1
-
8, 9
1337
AK302638.1
-
9, 10
1238
AY172659.1
-
7, 8
1029
AY354202.1
-
9, 10
1230
BC030688.2
-
9, 10
1388
4822550
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
immune response
serine-type peptidase activity
dipeptidyl-peptidase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0