Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123383.1 | NP_001116855.1 | 4 | 3284 | ||
NM_001123384.1 | NP_001116856.1 | 4 | 3284 | |||
NM_001123385.1 | NP_001116857.1 | 4 | 3213 | |||
NM_017745.5 | NP_060215.4 | 4 | 3213 | |||
XM_005272616.1 | XP_005272673.1 | 4 | 3345 | |||
XM_005272618.3 | XP_005272675.1 | 4 | 3011 | |||
XM_005272619.3 | XP_005272676.1 | 4 | 3671 | |||
XM_005272620.3 | XP_005272677.1 | 4 | 3671 | |||
XM_006724536.3 | XP_006724599.1 | 4 | 4447 | |||
XM_011543929.2 | XP_011542231.1 | 4 | 3121 | |||
XM_011543930.1 | XP_011542232.1 | 4 | 3134 | |||
XM_011543931.2 | XP_011542233.1 | 4 | 3244 | |||
XM_017029615.1 | XP_016885104.1 | 4 | 4447 | |||
XM_017029616.1 | XP_016885105.1 | 4 | 3671 | |||
GenBank mRNA | AB046795.2 | - | 3 | 2939 | ||
AF317391.1 | - | 4 | 3172 | |||
AF317392.1 | - | 4 | 3172 | |||
AK056538.1 | - | 4 | 3038 | |||
AK074286.1 | - | 1 | 2274 | |||
AY316592.1 | - | 4 | 3172 | |||
BC114220.1 | - | 4 | 3284 | 4384724 | ||
BC128387.1 | - | 2 | 2137 | |||
BC128456.1 | - | 3 | 2949 |