Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281932.1 | NP_001268861.1 | 19, 20 | 2418 | ||
NM_001281933.1 | NP_001268862.1 | 18, 19 | 2365 | |||
NM_017760.6 | NP_060230.5 | 18, 19 | 2365 | |||
NR_104054.1 | 21, 22 | 2576 | ||||
XM_005249547.2 | XP_005249604.1 | 18, 19 | 2363 | |||
XM_011516356.2 | XP_011514658.1 | 18, 19 | 2363 | |||
XM_011516357.2 | XP_011514659.1 | 19, 20 | 2391 | |||
XM_011516358.2 | XP_011514660.1 | 18, 19 | 2363 | |||
XM_011516362.1 | XP_011514664.1 | 6, 7 | 842 | |||
XM_017012374.1 | XP_016867863.1 | 18, 19 | 2363 | |||
XM_017012375.1 | XP_016867864.1 | 17, 18 | 2189 | |||
XM_017012376.1 | XP_016867865.1 | 17, 18 | 2189 | |||
XM_017012377.1 | XP_016867866.1 | 8, 9 | 1121 | |||
XM_017012378.1 | XP_016867867.1 | 15, 16 | 1768 | |||
GenBank mRNA | AK000318.1 | - | 13, 14 | 1623 | ||
AK022744.1 | - | 8, 9 | 1165 | |||
AK023347.1 | - | 7, 8 | 919 | |||
AK092008.1 | - | 19, 20 | 2353 | |||
AK295057.1 | - | 4, 5 | 584 | |||
AK301901.1 | - | 6, 7 | 843 | |||
AK315699.1 | - | 16, 17 | 1982 | |||
BC020560.1 | - | 13, 14 | 1689 | 3458938 | ||
BC043404.1 | - | 18, 19 | 2306 | 6045248 | ||
BX537845.1 | - | 18, 19 | 2332 |