Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017785.4 | NP_060255.3 | 9 | 1364 | ||
XM_011534585.1 | XP_011532887.1 | 9 | 1310 | |||
XM_011534586.2 | XP_011532888.1 | 9 | 1246 | |||
XM_011534587.2 | XP_011532889.1 | 10 | 1511 | |||
XM_011534588.2 | XP_011532890.1 | 10 | 1449 | |||
XM_011534589.2 | XP_011532891.1 | 9 | 1259 | |||
XM_011534590.2 | XP_011532892.1 | 8 | 1000 | |||
XM_017009601.1 | XP_016865090.1 | 9 | 1195 | |||
XM_017009602.1 | XP_016865091.1 | 8 | 1000 | |||
GenBank mRNA | AF269167.1 | - | 7 | 958 | ||
AK000371.1 | - | 8 | 1090 | |||
AK074227.1 | - | 6 | 769 | |||
AK304623.1 | - | 8 | 1028 | |||
AK310442.1 | - | 9 | 1163 | |||
AY517557.1 | - | 9 | 1195 | |||
BC012568.1 | - | 9 | 1230 | 4102558 | ||
BU166207.1 | - | 3 | 305 | |||
CD653505.1 | - | 1 | 13 |