Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017813.4 | NP_060283.3 | 5 | 1296 | ||
GenBank mRNA | AK000428.1 | - | 5 | 1293 | ||
AY032885.1 | - | 5 | 1010 | |||
BC017797.1 | - | 5 | 1245 | 4306100 | ||
BC067814.1 | - | 5 | 1269 | 5295820 |