Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDYHV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283024.1 | NP_001269953.1 | 5 | 542 | ||
NM_001283027.1 | NP_001269956.1 | 6 | 643 | |||
NM_018024.2 | NP_060494.1 | 5 | 618 | |||
NR_133926.1 | 5 | 542 | ||||
XM_006716597.3 | XP_006716660.1 | 5 | 612 | |||
XM_011517147.1 | XP_011515449.1 | 5 | 611 | |||
XM_011517151.2 | XP_011515453.1 | 6 | 710 | |||
XM_017013601.1 | XP_016869090.1 | 5 | 612 | |||
XM_017013602.1 | XP_016869091.1 | 5 | 535 | |||
XM_017013603.1 | XP_016869092.1 | 5 | 534 | |||
GenBank mRNA | AK001066.1 | - | 5 | 618 | ||
AK225643.1 | - | 5 | 569 | |||
AK293492.1 | - | 5 | 462 | |||
AL528914.3 | - | 5 | 534 | |||
AU119047.1 | - | 5 | 618 | |||
BC008781.2 | - | 5 | 536 | 3609961 | ||
BG528966.1 | - | 6 | 595 |