Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGOHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300739.2 | NP_001287668.1 | 4, 5 | 594 | ||
NM_001319985.1 | NP_001306914.1 | 4, 5 | 324 | |||
NM_018048.4 | NP_060518.1 | 4, 5 | 426 | |||
NR_135120.1 | 5, 6 | 712 | ||||
NR_135121.1 | 5, 6 | 610 | ||||
XM_005253402.1 | XP_005253459.1 | 5, 6 | 765 | |||
GenBank mRNA | AF165518.1 | - | 4, 5 | 422 | ||
AK001154.1 | - | 4, 5 | 411 | |||
AK022720.1 | - | 4, 5 | 401 | |||
AK096106.1 | - | 3, 4 | 2313 | |||
BC010905.1 | - | 4, 5 | 389 | 4283108 | ||
BF667243.1 | - | 4, 5 | 305 | |||
BG497574.1 | - | 3, 4 | 257 | |||
BG533740.1 | - | 4, 5 | 427 | |||
DB340467.1 | - | 1, 2 | 100 | |||
KF051004.1 | - | 4, 5 | 349 |