Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018141.3 | NP_060611.2 | 4 | 261 | ||
XM_005249206.2 | XP_005249263.1 | 4 | 270 | |||
XM_011514724.2 | XP_011513026.1 | 5 | 265 | |||
XM_011514725.2 | XP_011513027.1 | 5 | 257 | |||
XM_017010996.1 | XP_016866485.1 | 4 | 262 | |||
GenBank mRNA | AF113220.1 | - | 4 | 221 | ||
AF277180.1 | - | 4 | 223 | |||
AK001429.1 | - | 4 | 230 | |||
AK098446.1 | - | 4 | 224 | |||
AK298219.1 | - | 4 | 244 | |||
AK316599.1 | - | 4 | 227 | |||
BC012560.1 | - | 4 | 231 | 4277803 | ||
BG776014.1 | - | 4 | 262 |