Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142628.1 | NP_001136100.1 | 11 | 1427 | ||
NM_018144.3 | NP_060614.2 | 12 | 1493 | |||
GenBank mRNA | AF346603.1 | - | 12 | 1434 | ||
AK001440.1 | - | 5 | 728 | |||
AK022027.1 | - | 6 | 876 | |||
AK292295.1 | - | 12 | 1443 | |||
AK301841.1 | - | 11 | 1380 |