Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177663.1 | NP_001171134.1 | 7, 8 | 846 | ||
NM_001177664.1 | NP_001171135.1 | 7, 8 | 749 | |||
NM_001177665.1 | NP_001171136.1 | 6, 7 | 1019 | |||
NM_018151.4 | NP_060621.3 | 7, 8 | 846 | |||
XM_005246665.3 | XP_005246722.1 | 7, 8 | 726 | |||
XM_011511395.2 | XP_011509697.1 | 6, 7 | 952 | |||
XM_017004422.1 | XP_016859911.1 | 7, 8 | 819 | |||
XM_017004424.1 | XP_016859913.1 | 6, 7 | 663 | |||
GenBank mRNA | AK022932.1 | - | 2, 3 | 209 | ||
AK299291.1 | - | 7, 8 | 723 | |||
AY584066.1 | - | 6, 7 | 685 | |||
AY585745.1 | - | 7, 8 | 795 | |||
AY727910.1 | - | 7, 8 | 846 | |||
AY727911.1 | - | 7, 8 | 749 | |||
AY727912.1 | - | 6, 7 | 1019 | |||
AY727913.1 | - | 7, 8 | 846 | |||
BX648324.1 | - | 7, 8 | 815 | |||
CR933663.1 | - | 6, 7 | 659 |