Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCBTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018191.3 | NP_060661.3 | 4 | 409 | ||
XM_005266441.2 | XP_005266498.1 | 3 | 329 | |||
XM_011535133.1 | XP_011533435.1 | 4 | 435 | |||
XM_011535134.1 | XP_011533436.1 | 4 | 335 | |||
XM_011535135.2 | XP_011533437.1 | 3 | 244 | |||
XM_011535136.2 | XP_011533438.1 | 4 | 413 | |||
GenBank mRNA | AF334406.1 | - | 4 | 383 | ||
AJ319660.1 | - | 4 | 350 | |||
AK096654.1 | - | 4 | 409 | |||
AL833821.1 | - | 4 | 319 | |||
BC038104.2 | - | 4 | 403 | 5269720 |