Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282564.1 | NP_001269493.1 | 2 | 364 | ||
NM_018202.5 | NP_060672.2 | 2 | 364 | |||
XM_005245931.2 | XP_005245988.1 | 2 | 325 | |||
XM_011541704.2 | XP_011540006.1 | 2 | 318 | |||
GenBank mRNA | AK001609.1 | - | 2 | 275 | ||
AK311472.1 | - | 2 | 325 | |||
AY845015.1 | - | 2 | 294 | |||
AY845035.1 | - | 2 | 232 | |||
BC032427.1 | - | 2 | 232 | 5246967 | ||
DB455940.1 | - | 2 | 364 |