Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@6a7fd88a
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
See other ECHDC2 SIRNA Assays ›
Gene Symbol:
ECHDC2
Gene Name:
enoyl-CoA hydratase domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 52895910 - 52927212 on Build GRCh38
UniGene ID:
Hs.476319
Species:
Human
siRNA ID
HSS124260
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ECHDC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198961.1
NP_001185890.1
10
957
NM_001198962.1
NP_001185891.1
8
813
NM_001319958.1
NP_001306887.1
10
1377
NM_018281.3
NP_060751.2
9
864
XM_011541709.2
XP_011540011.1
12
1343
XM_011541710.2
XP_011540012.1
11
1168
XM_011541711.2
XP_011540013.1
11
1250
XM_011541712.2
XP_011540014.1
10
1120
XM_011541713.2
XP_011540015.1
11
5120
XM_011541715.2
XP_011540017.1
12
1568
XM_011541719.2
XP_011540021.1
12
2565
XM_011541720.2
XP_011540022.1
11
4889
XM_011541721.2
XP_011540023.1
13
1582
XM_011541722.2
XP_011540024.1
12
1200
XM_011541723.2
XP_011540025.1
12
1300
XM_011541724.2
XP_011540026.1
10
1193
XM_011541725.2
XP_011540027.1
10
1075
XM_011541726.2
XP_011540028.1
10
5027
XM_011541727.2
XP_011540029.1
9
4739
XM_017001638.1
XP_016857127.1
13
2954
XM_017001639.1
XP_016857128.1
12
1359
XM_017001640.1
XP_016857129.1
11
1522
XM_017001641.1
XP_016857130.1
13
2723
XM_017001642.1
XP_016857131.1
9
4852
GenBank mRNA
AF258590.1
-
5
1776
AK001810.1
-
9
816
AK302285.1
-
13
1536
AL832874.1
-
3
2025
BC038389.1
-
10
1295
BC044574.1
-
10
887
5314083
BC051887.1
-
8
2325
BX647186.1
-
1
5495
Back To Top
More Information
Panther Classification:
Molecular Function -
acetyltransferase
acyltransferase
dehydrogenase
epimerase/racemase
hydratase
isomerase
ligase
lyase
oxidoreductase
transferase
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-1-prod-blue:8080/10.253.225.14:8080.
git-commit: 578676318d66c763e4a01d7f975a961cd389eaca
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.18.0-2021.03.09-1.0