Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018290.3 | NP_060760.2 | 11, 12 | 1493 | ||
XM_011513711.2 | XP_011512013.1 | 12, 13 | 1596 | |||
GenBank mRNA | AF109360.1 | - | 11, 12 | 1433 | ||
AK001845.1 | - | 11, 12 | 1436 | |||
AK223237.1 | - | 11, 12 | 1435 | |||
AK297752.1 | - | 9, 10 | 1230 | |||
AL136705.1 | - | 11, 12 | 1450 | |||
BC010087.2 | - | 11, 12 | 1433 | 2964390 | ||
CR457274.1 | - | 11, 12 | 1420 |