Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161575.1 | NP_001155047.1 | 6 | 1167 | ||
NM_018296.5 | NP_060766.5 | 9 | 1437 | |||
XM_005256025.2 | XP_005256082.1 | 9 | 1437 | |||
XM_005256026.2 | XP_005256083.1 | 8 | 996 | |||
XM_005256027.2 | XP_005256084.1 | 9 | 1437 | |||
XM_005256028.1 | XP_005256085.1 | 8 | 1309 | |||
XM_011523200.1 | XP_011521502.1 | 9 | 1437 | |||
XM_011523201.1 | XP_011521503.1 | 6 | 1057 | |||
XM_011523202.2 | XP_011521504.1 | 5 | 912 | |||
XM_011523203.1 | XP_011521505.1 | 6 | 819 | |||
XM_011523204.1 | XP_011521506.1 | 7 | 1236 | |||
XM_011523205.1 | XP_011521507.1 | 8 | 1327 | |||
XM_011523206.2 | XP_011521508.1 | 9 | 1437 | |||
XM_017023400.1 | XP_016878889.1 | 9 | 1437 | |||
GenBank mRNA | AK001866.1 | - | 6 | 1216 | ||
AK292311.1 | - | 6 | 1146 | |||
AK297329.1 | - | 4 | 661 | |||
AK301766.1 | - | 6 | 1121 | |||
AK309897.1 | - | 5 | 709 | |||
AK311487.1 | - | 4 | 603 | |||
AY313780.1 | - | 6 | 1130 | |||
AY963267.1 | - | 9 | 1437 | |||
BC018031.1 | - | 8 | 999 | |||
BC026156.2 | - | 9 | 1437 | 4828487 |