Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161575.1 | NP_001155047.1 | 8 | 1510 | ||
NM_018296.5 | NP_060766.5 | 11 | 1780 | |||
XM_005256025.2 | XP_005256082.1 | 11 | 1780 | |||
XM_005256026.2 | XP_005256083.1 | 10 | 1339 | |||
XM_005256028.1 | XP_005256085.1 | 10 | 1652 | |||
XM_011523201.1 | XP_011521503.1 | 8 | 1400 | |||
XM_011523202.2 | XP_011521504.1 | 7 | 1255 | |||
XM_011523203.1 | XP_011521505.1 | 8 | 1162 | |||
XM_011523204.1 | XP_011521506.1 | 9 | 1579 | |||
XM_011523205.1 | XP_011521507.1 | 10 | 1670 | |||
XM_017023400.1 | XP_016878889.1 | 11 | 1780 | |||
GenBank mRNA | AK292311.1 | - | 8 | 1489 | ||
AY313780.1 | - | 8 | 1473 | |||
AY963267.1 | - | 11 | 1780 | |||
BC018031.1 | - | 10 | 1274 | |||
BC026156.2 | - | 11 | 1780 | 4828487 |