Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174098.1 | NP_001167569.1 | 6 | 926 | ||
NM_018344.5 | NP_060814.4 | 6 | 920 | |||
NR_033413.1 | 5 | 837 | ||||
NR_033414.1 | 4 | 610 | ||||
XM_006717910.3 | XP_006717973.1 | 6 | 1312 | |||
XM_017016377.1 | XP_016871866.1 | 5 | 1134 | |||
XM_017016378.1 | XP_016871867.1 | 4 | 910 | |||
GenBank mRNA | AF326987.1 | - | 5 | 868 | ||
AK002022.1 | - | 5 | 671 | |||
AK304503.1 | - | 5 | 807 | |||
AK314497.1 | - | 6 | 888 | |||
AK316152.1 | - | 5 | 822 | |||
AX775915.1 | - | 6 | 920 | |||
AX775917.1 | - | 6 | 920 | |||
AX775919.1 | - | 6 | 920 | |||
AX775921.1 | - | 6 | 920 | |||
AY288928.1 | - | 6 | 880 | |||
AY358686.1 | - | 6 | 898 | |||
BC000223.1 | - | 3 | 295 | |||
BC041575.1 | - | 5 | 808 | |||
BC120996.1 | - | 6 | 898 | 40116513 | ||
BC120997.1 | - | 6 | 898 | 40116514 |