Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGPAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018361.3 | NP_060831.2 | 8 | 1384 | ||
XM_011534751.2 | XP_011533053.1 | 6 | 904 | |||
GenBank mRNA | AF375789.1 | - | 8 | 1346 | ||
AK002072.1 | - | 8 | 1048 | |||
AK310545.1 | - | 3 | 523 | |||
AL136587.1 | - | 8 | 1074 | |||
BC023550.2 | - | 8 | 1328 | 4129973 | ||
BC068519.1 | - | 8 | 1048 | |||
BC080537.1 | - | 8 | 1353 | 5015128 |