Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127370.2 | NP_001120842.1 | 5 | 500 | ||
NM_001127371.2 | NP_001120843.1 | 3 | 308 | |||
NM_018719.4 | NP_061189.2 | 4 | 446 | |||
GenBank mRNA | AK022955.1 | - | 4 | 421 | ||
AK095965.1 | - | 5 | 454 | |||
AK292490.1 | - | 4 | 421 | |||
AK300781.1 | - | 3 | 268 | |||
AY161168.1 | - | 4 | 323 | |||
AY161169.1 | - | 4 | 326 | |||
BC009352.2 | - | 4 | 338 | 4139111 | ||
BC014630.2 | - | 3 | 290 | |||
BC025242.1 | - | 4 | 400 | 4842169 | ||
BC032576.1 | - | 4 | 411 | 5491780 | ||
BC039823.1 | - | 4 | 410 | 6057601 | ||
BX422040.2 | - | 3 | 256 | |||
DA041070.1 | - | 4 | 446 | |||
DC331792.1 | - | 3 | 308 |