Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017547.3 | NP_060017.1 | 10 | 1297 | ||
NR_037647.1 | 9 | 1076 | ||||
NR_037648.1 | 10 | 1316 | ||||
XM_006718879.3 | XP_006718942.1 | 9 | 1010 | |||
XM_006718881.3 | XP_006718944.1 | 8 | 978 | |||
XM_011542895.2 | XP_011541197.1 | 10 | 1394 | |||
XM_011542896.2 | XP_011541198.1 | 10 | 1203 | |||
XM_017018000.1 | XP_016873489.1 | 10 | 1269 | |||
XM_017018001.1 | XP_016873490.1 | 10 | 1416 | |||
XM_017018002.1 | XP_016873491.1 | 9 | 925 | |||
XM_017018003.1 | XP_016873492.1 | 9 | 1177 | |||
XM_017018004.1 | XP_016873493.1 | 9 | 1299 | |||
XM_017018005.1 | XP_016873494.1 | 8 | 2532 | |||
XM_017018006.1 | XP_016873495.1 | 9 | 1174 | |||
GenBank mRNA | AF103801.1 | - | 10 | 1195 | ||
AF447877.1 | - | 8 | 1449 | |||
AK023987.1 | - | 10 | 1166 | |||
AK126067.1 | - | 4 | 2275 | |||
AK295267.1 | - | 10 | 1252 | |||
AK295428.1 | - | 10 | 1182 | |||
AK298010.1 | - | 8 | 923 | |||
AK298807.1 | - | 10 | 1182 | |||
AK302039.1 | - | 9 | 945 | |||
AL136923.1 | - | 10 | 1177 | |||
BC002910.2 | - | 8 | 774 | |||
BC013902.2 | - | 10 | 1153 | 3956972 |