Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017619.3 | NP_060089.1 | 6, 7 | 744 | ||
GenBank mRNA | AK057799.1 | - | 6, 7 | 723 | ||
AK289844.1 | - | 6, 7 | 1009 | |||
AY099329.1 | - | 6, 7 | 729 | |||
BC010697.1 | - | 6, 7 | 720 | 3873751 |