Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017619.3 | NP_060089.1 | 7, 8 | 881 | ||
GenBank mRNA | AK000015.1 | - | 1, 2 | 56 | ||
AK057799.1 | - | 7, 8 | 860 | |||
AK289844.1 | - | 7, 8 | 1146 | |||
AL137730.1 | - | 1, 2 | 96 | |||
AY099329.1 | - | 7, 8 | 866 | |||
BC010697.1 | - | 7, 8 | 857 | 3873751 |