Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017631.5 | NP_060101.3 | 36 | 5251 | ||
XM_011532103.2 | XP_011530405.1 | 36 | 5251 | |||
GenBank mRNA | AK000042.1 | - | 8 | 1052 | ||
AK026333.1 | - | 4 | 517 | |||
AK223184.1 | - | 8 | 1052 | |||
AK225191.1 | - | 8 | 1039 | |||
AL137464.1 | - | 3 | 437 | |||
BC020601.1 | - | 4 | 467 | |||
BC038115.2 | - | 36 | 5196 | 5261001 | ||
BC074781.2 | - | 8 | 1030 | |||
BX648655.1 | - | 16 | 2093 |