Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF280C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017666.4 | NP_060136.1 | 13 | 1655 | ||
XM_006724765.3 | XP_006724828.1 | 12 | 1459 | |||
GenBank mRNA | AK000102.1 | - | 13 | 1578 | ||
AK290373.1 | - | 13 | 1606 | |||
AL133204.1 | - | 12 | 1443 | |||
AL834333.1 | - | 13 | 1586 | |||
BC051728.1 | - | 13 | 1606 | 4828435 |