Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308097.1 | NP_001295026.1 | 13 | 1928 | ||
NM_017769.4 | NP_060239.2 | 14 | 1969 | |||
XM_011536925.2 | XP_011535227.1 | 14 | 1935 | |||
XM_011536926.1 | XP_011535228.1 | 13 | 1852 | |||
GenBank mRNA | AB037754.1 | - | 14 | 1921 | ||
AK000340.1 | - | 14 | 1934 | |||
AK022867.1 | - | 4 | 744 | |||
AK225346.1 | - | 14 | 1934 | |||
AK296872.1 | - | 13 | 1893 | |||
BC000973.2 | - | 14 | 1915 | 3450488 | ||
BC040179.1 | - | 2 | 1225 |