Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017797.3 | NP_060267.2 | 7, 8 | 1259 | ||
XM_005259593.1 | XP_005259650.1 | 7, 8 | 1024 | |||
XM_011528127.1 | XP_011526429.1 | 5, 6 | 625 | |||
GenBank mRNA | AF355797.1 | - | 7, 8 | 1259 | ||
AK000393.1 | - | 4, 5 | 1053 | |||
AK027481.1 | - | 5, 6 | 621 | |||
AK056818.1 | - | 1, 2 | 1394 | |||
AK225831.1 | - | 7, 8 | 1258 | |||
AL360275.1 | - | 4, 5 | 490 | |||
AY007116.1 | - | 7, 8 | 981 | |||
BC000564.2 | - | 4, 5 | 549 | |||
BC008035.2 | - | 4, 5 | 543 | |||
BC053986.1 | - | 3, 4 | 444 | |||
BC069199.1 | - | 5, 6 | 685 | |||
BC094820.1 | - | 7, 8 | 993 | |||
CR457230.1 | - | 3, 4 | 296 |