Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIF1AN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017902.2 | NP_060372.2 | 7 | 1019 | ||
XM_011539940.2 | XP_011538242.1 | 7 | 1030 | |||
XM_011539941.1 | XP_011538243.1 | 7 | 897 | |||
GenBank mRNA | AF395830.1 | - | 7 | 954 | ||
AK000622.1 | - | 7 | 964 | |||
AK025680.1 | - | 3 | 178 | |||
AK295353.1 | - | 7 | 1046 | |||
AL359615.1 | - | 6 | 682 | |||
BC007719.2 | - | 7 | 947 | 4138066 |