Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018060.3 | NP_060530.3 | 17, 18 | 2269 | ||
GenBank mRNA | AK001188.1 | - | 15, 16 | 1744 | ||
AK001726.1 | - | 11, 12 | 1257 | |||
AK022665.1 | - | 7, 8 | 836 | |||
AK291432.1 | - | 17, 18 | 2078 | |||
AY267462.1 | - | 17, 18 | 2280 | |||
BC010218.1 | - | 13, 14 | 1449 | |||
BC040376.1 | - | 17, 18 | 2002 | |||
BC047880.1 | - | 16, 17 | 1796 | |||
BC137438.1 | - | 17, 18 | 2239 | 9021436 | ||
D28500.1 | - | 17, 18 | 2110 |